Learn More
Medchemexpress LLC PG-11047 tetrahydrochloride | 206991-64-2 | 98.0% | 402.27 g/mol | C14H36Cl4N4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1639410MG
Encompass_Preferred
PG-11047 (CGC-11047) is a conformationally restricted polyamine analogue supplied as the tetrahydrochloride salt for research use. It functions as a nonfunctional competitor of the natural polyamine spermine and has demonstrated broad-spectrum anticancer activity in preclinical and early clinical studies. Provided as a 10 MG solid for laboratory research.
  • Acts as a nonfunctional competitor of spermine.
  • Demonstrates broad-spectrum anticancer activity in preclinical and early clinical studies.
  • Supplied as the tetrahydrochloride salt in solid form.
  • Reported purity greater than 98.0%.
  • Molecular formula C14H36Cl4N4; molecular weight 402.27 g/mol.
  • Intended for research use only; not for therapeutic use.
Catalog No. 50-002-89127