Learn More
Medchemexpress LLC PG01037 dihydrochloride | 675599-62-9 | MFCD20527309 | 99.4% | 554.34 g/mol | C26H28Cl4N4O | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1034081MG
Encompass_Preferred
PG01037 dihydrochloride is a research-grade dopamine D3 receptor antagonist with high potency (Ki = 0.7 nM). It is supplied as a solid powder for use in receptor pharmacology, selectivity profiling, and in vitro or in vivo studies, with solubility in DMSO and reported high purity.
Catalog No. 50-002-44912