Learn More
Cygnus Technologies PG13 HOST CELL PROTEIN ELISA
Supplier:  Cygnus Technologies F995
Encompass
PG13 Host Cell Protein ELISA Kit PG13 cell line is used for cost-effective production of a high-titer retrovirus capable of infecting human cells for gene and cell therapies. The manufacturing and purification process of these products leaves the potential for PG13-derived host cell protein (HCP) impurities. It is required to reduce HCP impurities to the lowest levels practical.
Catalog No. NC2284877