Learn More
Medchemexpress LLC Pgam1-in-2 | 2438637-66-0 | 98.0% | 488.47 g/mol | C25H16N2O7S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1286825MG
Encompass_Preferred
PGAM1-IN-2 is a small-molecule inhibitor of phosphoglycerate mutase 1 (PGAM1) developed for biochemical and cellular research. It inhibits PGAM1 with an IC50 of 2.1 μM, has a molecular weight of 488.47 g/mol, and is provided as a high-purity solid or as a 10 mM solution in DMSO for convenient use in assays.
  • Potent PGAM1 inhibition (IC50 2.1 μM).
  • High purity suitable for research applications (98.0%).
  • Available as solid and as a 10 mM DMSO solution for assay flexibility.
  • Supplied in multiple small-scale pack sizes for screening and follow-up studies.
  • Molecular weight and formula provided for analytical and preparation needs.
Catalog No. 50-002-62513