Learn More
Medchemexpress LLC PGAM1-IN-2 | 2438637-66-0 | 98.0% | 50 MG

Supplier: Medchemexpress LLC HY12868250MG
PGAM1-IN-2 is an inhibitor of phosphoglycerate mutase 1 (PGAM1) with an IC50 of 2.1 μM. It inhibits H1299 proliferation with an IC50 of 33.8 ± 6.1 μM. This compound has a molecular weight of 488.47 and a chemical formula of C25H16N2O7S. It appears as a solid, light yellow to yellow in color.
- Inhibitor of phosphoglycerate mutase 1 (PGAM1)
- IC50 of 2.1 μM against PGAM1
- Inhibits H1299 proliferation with an IC50 of 33.8 ± 6.1 μM
- Molecular weight: 488.47
- Chemical formula: C25H16N2O7S
- Appearance: Solid, light yellow to yellow
- Solubility in DMSO: 62.5 mg/mL (127.95 mM)
- Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
- Recommended storage for in solvent: -80°C for 6 months, -20°C for 1 month
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.