Learn More
Medchemexpress LLC PGE-M 25ug | 328.36 | 25UG
SDP
Supplier:  Medchemexpress LLC HY11498825UG
Encompass_Preferred
PGE-M is a metabolite of prostaglandin E2 (PEG2) as a biomarker of inflammation and cancer including advanced colorectal neoplasia ovarian cancer prostate cancer and so on Urinary PGE-M is positively associated with obesity smoking and lung metastases with breast cancer[1][2][3]
Catalog No. 50-002-29594