Learn More
TARGETMOL CHEMICALS INC PGP-4008 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T283965MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. PGP-4008 is a selective and potent P-glycoprotein (Pgp) inhibitor that inhibits tumor growth by co-administration with Doxorubicin. purity: 98%
Catalog No. 50-258-0520