Learn More
Medchemexpress LLC HY-101587 5mg Medchemexpress, PGS-IN-1 CAS:102271-49-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1015875MG
Encompass_Preferred
Medchemexpress, HY-101587 5mg PGS-IN-1 CAS:102271-49-8 PGS-IN-1 is a potent inhibitor of prostaglandin synthetase (PGS) with an IC 50 of 0.28 μM; also inhibits 5-lipoxygenase with an IC 50 of 1.05 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2180