Learn More
YMC America, Inc Ph 120A 3um 75x0.3mm 1/16
SDP
Supplier:  YMC America, Inc PH12S03L5H0AU
Encompass
Phenyl columns provide unique selectivity when compared to aliphatic straight chain reversed phase materials such as C18 C8 and C4 YMC-Pack Phenyl is a monomerically-bonded phenyl and is fully endcapped Typical applications include aromatic compounds phenols fullerenes sweeteners
Catalog No. 50-008-78238