Learn More
HANNA INSTRUMENTS INC pH 4.01 Calibration Buffer (25 x 20 mL) Sachets
SDP
Supplier:  HANNA INSTRUMENTS INC HI70004C
Encompass
HI70004C is a premium quality pH 4.01 calibration buffer that is NIST traceable and supplied with a Certificate of Analysis. Each sachet has the lot number and expiration date stamped on it and is made of light block foil ensuring freshness each time one is opened. Hanna's line of technical calibration buffers have been specially formulated to have an expiration of 5 years from the date of manufacture for an unopened sachet. The HI70004C is for 25 sachets containing 20 mL of buffer.Supplied with Certificate of AnalysisAccuracy of +/- 0.01 pH @ 25°CTemperature chart of the actual pH value at various temperatures printed on each sachet
Catalog No. NC1865822