Learn More
Waters Corp pH Buffer Solution, pH 4, 1 Pint, NIST Traceable
Supplier:  Waters Corp 127
Encompass_Preferred
This pH 4 buffer comes in a 1 Pint bottle. It is directly traceable to NIST SRMs, mercury free, guaranteed stable for at least one year after receipt, and supplied with a full certificate of analysis.
  • Directly traceable to NIST SRMs
  • Mercury free
  • Guaranteed stable for at least one year after receipt
  • Supplied with a full certificate of analysis
Catalog No. 50-465-877