Learn More
Andwin Scientific PH 7 BUFFER SOLUTION, 4L POUCH
SDP
Supplier:  Andwin Scientific BDH50504L
Encompass_Preferred
The BDH pH reference standard buffer is a clear, yellow, odorless, liquid solution. This contains hydrochloric acid, potassium chloride, and deionized water. It is classified to have a 6.99-7.01 pH at 25.0 �C. This product is packaged and sold in a 4 L cubitainer.
Catalog No. NC0389126