Learn More
Waters Corp Waters pH Buffer, pH 7, 1 Liter, NIST Traceable
Supplier:  Waters Corp 133
Encompass_Preferred
This pH Buffer is supplied in a 1 Liter bottle, offering direct traceability to NIST SRMs. It is mercury free, guaranteed stable for at least one year after receipt, and comes with a full certificate of analysis.
  • Directly traceable to NIST SRMs.
  • Mercury free.
  • Guaranteed stable for at least one year after receipt.
  • Supplied with a full certificate of analysis.
  • Liquid formulation.
  • Designed for room temperature storage.
  • Has a 12-month shelf life.
  • Suitable for calibration applications.
  • Conforms to EPA method 150.1.
Catalog No. 50-465-880