Learn More
Medchemexpress LLC PH-797804 10mg | 586379-66-0 | 10MG
SDP
Supplier:  Medchemexpress LLC HY1040310MG
Encompass_Preferred
PH-797804 is a ATP-competitive selective p38 /p38 inhibitor (IC50 26 nM and Ki 5 8 nM for p38 Ki 40 nM for p38 ) and does not inhibit JNK2
Catalog No. 50-002-41915