Learn More
Selleck Chemical LLC PH-797804 5mg 586379-66-0
SDP
Supplier:  Selleck Chemical LLC S27265MG
Encompass_Preferred
PH-797804 is a novel pyridinone inhibitor of p38? with IC50 of 26 nM in a cell-free assay; 4-fold more selective versus p38? and does not inhibit JNK2. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0283482