Learn More
Eurofins Discoverx PathHunter Bioassay Detection Kit, 500 (10,000 dp, 96-well). (93-0933L)
Supplier:  Eurofins Discoverx 930933L
Encompass
The PathHunter Bioassay Detection Kit is a detection kit intended for use with Eurofins DiscoverX's PathHunter Bioassay cells or cell lines. This kit contains lysis buffer and EFC substrate to allow for detection of EFC levels in the assayed cells. This kit contains all components required to assay 100 plates (96-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-93-0933l.
Catalog No. 50-200-1600
Add to Cart
Request a Quote