Learn More
Hach Company Buffer Solution, pH 1.09, 500 mL
Supplier:  Hach Company S11M009
Encompass
pH Buffer Calibration reagent. Observed shelf-life after opening, following recommended storage and use: 3 months.
Catalog No. NC1626618