Learn More
Cole-Parmer Oakton Traceable One-Shot™ Buffer Solution, Red, pH 4.005; 6 x 100 mL Vials
Supplier:  Cole-Parmer 9876777
Encompass_Preferred
All Oakton Traceable pH buffer solutions are NIST-traceable reference materials Supplied with calibration report from an ISO 17034 and ISO 17025 accredited laboratory Individually packed in foil pouch to prevent UV exposure that may degrade accuracy Buffer solution lets you calibrate pH instruments to ensure accuracy If unopened, shelf life is 2 years. Convenient, single-use vials eliminate concerns of cross-contamination; each pack contains six polyethylene vials with 100-mL of pH buffer per vial. If unopened, shelf life is 2 years; once opened, use pH 4 and pH 7 within 90 days, pH 10 within 30 days Oakton Traceable One-Shot™ pH Buffer Solutions are supplied with NIST-traceable calibration reports stating accuracy at 25C to 0.010 pH, in addition to conversion values for pH/temperaturefor the most accurate calibrations!
Catalog No. 50-196-1816