Learn More
Hach Company pH Encap Sens-PPS-5 Wr-Convert
Supplier:  Hach Company 6022P050
Encompass
pH Encapsulated Sensor-PPS Body-5 Wire-Convertible-50-ft CablePPS Convertible Mounting Style 15m Analog Cable 5-Wire with Built-In Preamplifier General Purpose Glass pH Electrode
Catalog No. 50-002-07429