Learn More
YMC America, Inc Ph-HG col 120A 10um 250x10mm
SDP
Supplier:  YMC America, Inc PHG12S112510WT
Encompass_Preferred
Phenyl columns provide unique selectivity when compared to aliphatic straight chain reversed phase materials such as C18 C8 and C4 YMC-Pack Phenyl is a monomerically-bonded phenyl and is fully endcapped Typical applications include aromatic compounds phenols fullerenes sweeteners
Catalog No. 50-008-80391