Learn More
Hach Company Buffer Solution, pH 10.01, 125 mL
Supplier:  Hach Company LZW9470.99
Encompass
pH Buffer Standard Solution for pH electrode calibration, Colorless pH buffer, pH is 10.01 ±0.02 at 25°C, 125 mL bottle
Catalog No. 50-199-8011