Learn More
Medchemexpress LLC PHA 408 | 503555-55-3 | 99.4% | C29H27ClFN7O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY141801MG
Encompass_Preferred
PHA 408 is a potent, selective, and orally active IκB kinase-2 (IKK-2) inhibitor. It is a powerful anti-inflammatory agent against lipopolysaccharide (LPS)- and cigarette smoke (CS)-mediated lung inflammation.
  • Potent, selective, and orally active inhibitor
  • Powerful anti-inflammatory agent
  • Effective against LPS-induced lung inflammation
  • Effective against cigarette smoke-induced lung inflammation
Catalog No. 50-003-53427