Learn More
Selleck Chemical LLC PHA-665752 5mg 477575-56-7
SDP
Supplier:  Selleck Chemical LLC S10705MG
Encompass_Preferred
PHA-665752 is a potent, selective and ATP-competitive c-Met inhibitor with IC50 of 9 nM in cell-free assays, >50-fold selectivity for c-Met than RTKs or STKs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0687097