Learn More
TARGETMOL CHEMICALS INC PHA-680632 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T633825MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. PHA-680632 is potent inhibitor of Aurora A, Aurora B and Aurora C with IC50 of 27 nM, 135 nM and 120 nM, respectively. It has 10- to 200-fold higher IC50 for FGFR1, FLT3, LCK, PLK1, STLK2, and VEGFR2/3. Purity 98.2%
Catalog No. 50-287-6869