Learn More
Selleck Chemical LLC PHA-767491 HCl S2742-10mg
SDP
Supplier:  Selleck Chemical LLC S274210MG
Encompass_Preferred
PHA-767491 (CAY10572 NMS 1116354) HCl is a potent ATP-competitive dual Cdc7/CDK9 inhibitor with IC50 of 10 nM and 34 nM in cell-free assays respectively It displays 20-fold selectivity against CDK1/2 and GSK3- 50-fold selectivity against MK2 and CDK5 100-fold selectivity against PLK1 and CHK2
Catalog No. 50-310-8099