Learn More
Apexbio Technology LLC PHA-793887, 25mg. Cas: 718630-59-2 MFCD: MFCD17169992. Pan-Cdk inhibitor
SDP
Supplier:  Apexbio Technology LLC A545925
Encompass_Preferred
Description:IC50: PHA-793887 is a novel and potent inhibitor of CDK2, CDK5 and CDK7 with IC50 values of 8 nM, 5 nM and 10 nM, respectively. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4952