Learn More
Aat Bioquest Phalloidin Amine
SDP
Supplier:  Aat Bioquest 5302
Encompass_Preferred
Phalloidin, a bicyclic heptapeptide toxin, binds specifically at the interface between F-actin subunits, locking adjacent subunits together.
Catalog No. 50-195-3283