Learn More
Biotium Phalloidin, CF680, 50 U
Supplier:  Biotium 00053T
Encompass_Preferred
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red/ near-IR fluorescent CF680 has excitation/emission at 681/698 nm.
Catalog No. 50-196-4079