Learn More
Saint Gobain Performance Plastics PHARM BPT PUMP TUBING 25FT/PK
Supplier:  Saint Gobain Performance Plastics AY242038
Encompass_Preferred
Saint-Gobain PharMed BPT Tubing Diameter Inner 1/2 in. 12.7 mm Diameter Outer 3/4 in. 19 mm Thickness 0.13 in. 3.2 mm Autoclavable Autoclavable Certifications/Compliance Fully tested to ISO 10993 standards. Meets USP Class VI FDA and NSF criteria.
Catalog No. 50-206-9043