Learn More
AVANTOR MASTERFLEX (VWR LLC) Dow Corning Pharma-65 Silicone Tubing, 1/2"" x 3/4""; 50 ft/Pk
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9610711
Encompass
Applications: Pharmaceutical, biotechnology, and ultra-pure fluid processing. Characteristics: Platinum-cured silicone tubing with a very low extractable profile; it contains no phthalates, plasticizers, latex, or other organic additives. Remains flexible and resilient for long service life. Dow Corning maintains complete traceability from silicon to silicone tubing. Durometer shore A rating of 62. Certification: Elastomer is FDA compliant and meets USP Class VI requirements. Meets 3A Sanitary Standards and European Pharmacopoeia (EP) requirements. Tubing is manufactured according to Bulk Pharmaceutical (BP) and current GMP. Temperature range: ¿58 to 482¿F (¿50 to 250¿C).
Catalog No. NC1753597