Learn More
Sigma Aldrich Fine Chemicals Biosciences PharmalyteR 8 10.5 Cytiv
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17045501
Encompass
Pharmalyte(R) carrier ampholytes prepared by the co-polymerization of glycine glycylglycine amines and epichlorohydrin are available in five broad-range and four narrow-range pH intervals. Each interval contains numerous ampholytes with a high buffering capacity per pH unit. Pharmalyte(R) carrier ampholytes form an extremely stable linear pH gradient and exhibit even conductivity across the gel. For precast PhastGel(R) gels for PhastSystem see PhastGel.
Catalog No. 50-290-8403