Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuflow Systems Saint-Gobain PharmaPure Peristaltic Tubing, 3/8 in, 25 ft

Supplier:  Accuflow Systems AL242029

Encompass_Preferred

Saint-Gobain PharmaPure low-spallation peristaltic pump tubing is engineered for pharmaceutical, biotechnology, and laboratory fluid transfer. It provides extended pump life with ultra-low particulate spallation and low gas permeability to protect sensitive fluids and reduce contamination risk.

  • Low spallation peristaltic tubing designed for long pump life.
  • 3/8 in inner diameter.
  • 5/8 in outer diameter, 1/8 in wall thickness.
  • 25 ft length, opaque white color.
  • Maximum pressure: 19 psi at 23°C, 10 psi at 82°C.
  • Sterilization: autoclavable; ethylene oxide; gamma irradiation (25 kGy).
  • Typical applications: cell harvest, media processing, bioreactor process lines, aseptic filling.

Catalog No. NC2861983


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.