Learn More
Accuflow Systems Saint-Gobain PharmaPure Tubing, Thermoplastic Elastomer, 3/16 in x 5/16 in
Supplier:  Accuflow Systems AL242012
Encompass_Preferred
PharmaPure Low Spallation Peristaltic Pump Tubing from Saint-Gobain is a thermoplastic elastomer tubing designed for peristaltic pump applications that require minimal particulate generation, low gas permeability, and extended flex life. It is intended for bioprocessing, aseptic filling, and pharmaceutical fluid handling where product integrity is critical.
  • Thermoplastic elastomer material with low spallation and long flex life.
  • 3/16 in inner diameter, 5/16 in outer diameter, supplied as 25 ft coil.
  • Low gas permeability and reduced absorption for sensitive fluids.
  • Withstands repeated autoclaving and common sterilization processes.
  • Meets USP <88> Class VI, USP <87>, and ISO 10993-5 biocompatibility criteria.
  • Optimized for peristaltic pumps to reduce downtime and contamination risk.
Catalog No. 50-236-5829