Learn More
Accuflow Systems Pharmed 06508-92 tubing has dimensions of 1" ID x 1.375" OD.
Supplier:  Accuflow Systems 0650892
Encompass_Preferred
PharMed Tubing with an Inside Diameter of 1 in. and an Outside Diameter of 1-3/8 in. Wall thickness is 0.188 in. Pharmed tubing can be sterilized either by Autoclave or Gamma Irradiation, Certifications/Compliance Fully tested to ISO 10993 standards. Meets USP Class VI FDA and NSF criteria.
Catalog No. NC2162244