Learn More
Cole-Parmer PharMed BPT Biocompatible Peristaltic Pump Tubing, 3/4" ID x 1" OD, 25 Ft; Saint Gobain AY242053
Supplier:  Cole-Parmer 5012365
Encompass_Preferred
Biocompatible inner bore is perfect for sensitive bioprocess fluids
  • USP Class VI and USP <87> compliant
  • 3/4" ID x 1" OD
  • The biocompatible fluid surface and exceptional flex life makes this tubing ideal for transferring sensitive bioprocessing fluids in cell harvesting, media processing, vaccine manufacturing, aseptic filling, and diagnostic testing applications.
    The PharMed BPT tubing withstands repeated CIP and SIP, simplifying cleaning and sterilization. Tubing can be used with most commercial cleaners and sanitizers; autoclave up to five cycles or use gamma irradiation without affecting overall service life.
    Tubing features exceptional flex life characteristics and wear properties to outlast silicone in peristaltic pumps and help minimize downtime due to tubing failures. Low permeability characteristics provide protection for sensitive fluids from gas ingress. Opaque tubing protects light-sensitive fluids from visible and UV lights.
    Catalog No. 50-006-10277