Learn More
Accuflow Systems PharMed BPT Tubing, 1/32 in ID, 5/32 in OD
Supplier:  Accuflow Systems AY242606
Encompass_Preferred
PharMed BPT tubing is a biocompatible peristaltic pump tubing formulated for extended flex life and reduced gas and vapor permeability compared with silicone. It is intended for diagnostic, laboratory, and bioprocess fluid transfer where chemical resistance and biocompatibility are required.
  • 1/32 in inner diameter, 5/32 in outer diameter.
  • Standard 25 ft (7.6 m) spool length.
  • Maximum service temperature 275°F (135°C); low temperature embrittlement -75°F (-59°C).
  • Meets USP <88> Class VI and references to ISO 10993-5.
  • Very good general chemical resistance; excellent resistance to acids, alkalis, and oxidizers.
  • Formulated for peristaltic pump use with extended flex life and reduced gas permeability compared with silicone.
  • Typical applications include peristaltic pump lines, cell harvest and media transfer, bioreactor process lines, aseptic filling, and diagnostics.
Catalog No. 50-236-5823