Learn More
Saint Gobain Performance Plastics PHARMED BPT PUMP TUBING
Supplier:  Saint Gobain Performance Plastics AY242007
Encompass_Preferred
Saint-Gobain PharMed BPT Tubing Diameter Inner 0.13 in. 3.2 mm Diameter Outer 1/4 in. 6.4 mm Thickness 0.063 in. 1.6 mm Autoclavable Autoclavable Certifications/Compliance Fully tested to ISO 10993 standards. Meets USP Class VI FDA and NSF criteria.
Catalog No. 50-206-9036