Learn More
Sigma Aldrich Fine Chemicals Biosciences Pharmed L/S tubing size 15
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T069325FT
Encompass
Tubing for Masterflex Peristaltic Pumps PharMed tubing has been specifically formulated for the Masterflex pump system. Great for tissue and cell work- nontoxic and nonhemolytic. Long service life minimizes risk of fluid exposure reduces tubing cost and pump downtime.. Opaque to UV and visible light to protect light-sensitive fluids. Heat sealable bondable and formable. Extremely low gas permeability. Meets USP Class VI fDA 21 CfR 177.2600 and NSf listed (Standard 51)
Catalog No. 50-290-0041