Learn More
Bio Rad Laboratories PHARMED TUBING
Supplier:  Bio Rad Laboratories 7318209
Encompass
Tubingm PharMed; Bio-Rad; Wider range of chemical compatibility; Ideal tubing material for use in pumpheads of Model EP-1 Econo pump and BioLogic LP system; Autoclavable; 3.2mm I.D.; 1.0mm wall; 10m
Catalog No. 50-901-0191