Learn More
Medchemexpress LLC Phaseic acid | 24394-14-7 | 98.8% | 280.32 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1119731MG
Encompass_Preferred
Phaseic acid is an Abscisic acid terpenoid catabolite that can activate a subset of Abscisic acid receptors. It is a plant hormone associated with photosynthesis arrest and abscission. It also acts as an antagonist for NMDA-type glutamate receptor (NMDAR) and exhibits a neuroprotective effect.
  • Activates a subset of Abscisic acid receptors
  • Inhibits NMDAR currents with an IC50 of 34.37 μM
  • Reduces intracellular calcium influx
  • Exhibits a neuroprotective effect
  • Ameliorates ischemic brain lesions in MCAO models
Catalog No. 50-004-39013