Learn More
Medchemexpress LLC Phaseic acid | 24394-14-7 | MFCD00191183 | 98.8% | 280.32 g/mol | C15H20O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1119735MG
Encompass_Preferred
Phaseic acid is a terpenoid catabolite of the plant hormone abscisic acid used as a research-grade small molecule for biochemical and pharmacological studies. It has reported activity at abscisic acid receptors and shows antagonist activity at NMDA-type glutamate receptors, and is supplied as a purified solid for laboratory assays.
  • High purity solid suitable for research assays.
  • Interacts with abscisic acid receptors in biochemical studies.
  • Demonstrated NMDA receptor antagonist activity in pharmacology assays.
  • Provided in accurately measured small quantities for experimental work.
  • Molecular formula C15H20O5; molecular weight 280.32 g/mol.
Catalog No. 50-002-64708