Learn More
Medchemexpress LLC Phaseollin | 13401-40-6 | 1 MG

Supplier: Medchemexpress LLC HYN30871MG
Phaseollin is an isoflavonoid phytoalexin that can be isolated from Phaseolus vulgaris. Phaseollin is toxic to bean hypocotyl and endocarp cells, and causes a complete lysis of sheep erythrocytes.
- Purity of 98.0%
- Molecular weight of 322.35
- Molecular formula C20H18O4
- Solid appearance
- White to off-white color
- Store at -20°C, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.