Learn More
Medchemexpress LLC HY-N7400 5mg Medchemexpress, Phaseoloidin CAS:118555-82-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN74005MG
Encompass_Preferred
Medchemexpress, HY-N7400 5mg Phaseoloidin CAS:118555-82-1 Phaseoloidin is a homogentisic acid glucoside from Nicotiana attenuata trichomes and contributes to the plant's resistance against lepidopteran herbivores. Phaseoloidin reduces larval growth of the specialist larvae Manduca sexta and the generalist larvae Spodoptera littoralis. Phaseoloidin has anti-complement activitie. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2996