Learn More
Medchemexpress LLC Phe-Arg-β-naphthylamide dihydrochloride | 100929-99-5 | MFCD00058046 | 99.9% | 519.47 | C25H32Cl2N6O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY101444A5MG
Encompass_Preferred
PAβN dihydrochloride is a research-grade efflux pump inhibitor used to block bacterial multidrug efflux pumps and increase antibiotic susceptibility in Gram-negative bacteria. Supplied as the dihydrochloride salt, it is intended for biochemical and microbiology assays.
  • Inhibits bacterial multidrug efflux pumps to sensitize bacteria to antibiotics.
  • Suitable for in vitro antibacterial and efflux studies.
  • High purity suitable for analytical and research applications.
  • Supplied in small, precise quantities for experimental use.
  • Stable as a solid for standard laboratory storage and handling.
Catalog No. 50-002-49284