Learn More
Medchemexpress LLC Phe-Arg-β-naphthylamide dihydrochloride | 100929-99-5 | MFCD00058046 | 99.9% | 519.47 | C25H32Cl2N6O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY101444A50MG
Encompass_Preferred
PAβN dihydrochloride is the dihydrochloride salt of Phe-Arg-β-naphthylamide used as a research reagent to inhibit bacterial efflux pumps and study multidrug resistance mechanisms in Gram-negative organisms. Supplied as a high-purity solid for laboratory use (research use only).
  • Research-grade efflux pump inhibitor for Gram-negative bacteria.
  • Dihydrochloride salt form for consistent handling and formulation.
  • High purity suitable for biochemical and microbiological assays (~99.9%).
  • Molecular weight 519.47 g·mol⁻¹ and formula C25H32Cl2N6O2.
  • Available in small research pack sizes such as 50 MG.
Catalog No. 50-002-45810