Learn More
Carbosynth LLC. Phebestin (5 mg vial), 1ea, MW: 441.53, Molecular formula: C24H31N3O5, CAS# 187402-73-9
SDP
Supplier:  Carbosynth LLC. IPH4342V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: (2S,3R)-3-Amino-2-Hydroxy-4-Phenylbutanoyl-L-Valyl-L-Phenylalanine, Application: Synthetic Product Inhibitor for Aminopeptidase, Storage Temperature: -20C, References: N M. Nagai, F. Kojima, H. Naganawa, M. Hamada, T. Aoyagi, and T. Takeuchi, J. Antibiotics, 50, 82 (1997). (Original)
Catalog No. 50-168-5893