Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-Phellandrene | 99-83-2 | 86.7% | 10 G
SDP

Supplier:  Medchemexpress LLC HY12161510G

Encompass_Preferred

α-Phellandrene (CAS 99-83-2) is a cyclic monoterpene (C10H16) found in plant essential oils and used as a research reagent. It has reported insecticidal activity and biological effects in apoptosis, autophagy, cAMP signaling, and inflammatory/cancer models.

  • Cyclic monoterpene with molecular formula C10H16 and molecular weight 136.23 g/mol.
  • Reported insecticidal and anti-inflammatory activities useful for biological studies.
  • Used in research on apoptosis, autophagy, and NO/cAMP signaling pathways.
  • Storage guidance: pure form -20°C (3 years), 4°C (2 years); in solvent -80°C (6 months), -20°C (1 month).
  • Available in small laboratory pack sizes suitable for research use.

Catalog No. 50-002-30963


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.