Learn More
Medchemexpress LLC Phen-2,3,5,6-d4-ol-d, 4,4'-[1-(methyl-d3)ethylidene-2,2,2-d3]bis- | 96210-87-6 | 99.95% | 244.38 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY18260S50MG
Encompass_Preferred
Bisphenol A-d16 is the deuterium labeled Bisphenol A. Bisphenol A is a phenolic, organic synthetic compound widely used in the production of polycarbonate plastics and epoxy resins. It is classified as a reproductive, developmental, and systemic toxicant, and often as an endocrine-disrupting compound (EDC). Bisphenol A is linked to various diseases, including cardiovascular diseases, respiratory diseases, diabetes, kidney diseases, obesity, and reproductive disorders.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-87623