Learn More
Selleck Chemical LLC Phen-DC3 Triflate-E1070-5MG
SDP
Supplier:  Selleck Chemical LLC E10705MG
Encompass_Preferred
Phen-DC3 Trifluoromethanesulfonate is a G-quadruplex (G4) specific ligand which can inhibit FANCJ and DinG helicases with IC50s of 65 6 and 50 10 nM respectively
Catalog No. 50-313-6522