Learn More
Medchemexpress LLC HY-15594A 1mg Medchemexpress, Phen-DC3 Trifluoromethanesulfonate CAS:929895-45-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15594A1MG
Encompass_Preferred
Medchemexpress, HY-15594A 1mg Phen-DC3 Trifluoromethanesulfonate CAS:929895-45-4 Phen-DC3 Trifluoromethanesulfonate is a G-quadruplex (G4) specific ligand which can inhibit FANCJ and DinG helicases with IC50s of 65±6 and 50±10 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9145